Learn More
Electron Microscopy Sciences XYZ-Axis Electrically Conductive Tape 9712, 1/2" wide 36 YD
SDP
Catalog No. 50364055
Encompass_Preferred
Conductive Tape 9712 is an isotropically conductive pressure sensitive tape. The tape consists of a 3M adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance with very high conductivity. The 9712 Tape is an excellent choice when needing to bond your samples for all SEM work. Based upon four wire (Kelvin probe) resistance measurements made with crossed pieces of Foil/Type 9712 or 9713/Rigid plate construction using a 1.0" x 1.0" square piece of 3M tape Type 9712 or 9713. The rigid metal surface was prepared with a Scotch-Brite pad to roughen the surface and cleaned with isopropyl alcohol. Comes pn a 3" (76 mm) core.
Width: ½"
Catalog No. 50-364-055 Supplier Electron Microscopy Sciences Supplier No. 7780812
Conductive Tape 9712 is an isotropically conductive pressure sensitive tape. The tape consists of a 3M adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance with very high conductivity. The 9712 Tape is an excellent choice when needing to bond your samples for all SEM work. Based upon four wire (Kelvin probe) resistance measurements made with crossed pieces of Foil/Type 9712 or 9713/Rigid plate construction using a 1.0" x 1.0" square piece of 3M tape Type 9712 or 9713. The rigid metal surface was prepared with a Scotch-Brite pad to roughen the surface and cleaned with isopropyl alcohol. Comes pn a 3" (76 mm) core.
Width: ½"