Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y-25130 hydrochloride, Tocris Bioscience™

Catalog No. 038050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-805-0 50 mg
03-801-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 03-805-0 Supplier R&D Systems Supplier No. 0380/50
This item is not returnable. View return policy

Potent and selective 5-HT3 antagonist

A selective and potent 5-HT3 antagonist (Ki = 2.9 nM).

Chemical Identifiers

CAS 123040-16-4
Molecular Formula C17H21Cl2N3O3
Molecular Weight (g/mol) 386.273
InChI Key DBMKBKPJYAHLQP-UHFFFAOYSA-N
Synonym azasetron hydrochloride, azasetron hcl, serotone, y-25130 hydrochloride, c17h20cln3o3.hcl, azasetron hydrochloride jan, 2h-1,4-benzoxazine-8-carboxamide, 3,4-dihydro-n-1-azabicyclo 2.2.2 oct-3-yl-6-chloro-4-methyl-3-oxo-, monohydrochloride, 2h-1,4-benzoxazine-8-carboxamide, n-1-azabicyclo 2.2.2 oct-3-yl-6-chloro-3,4-dihydro-4-methyl-3-oxo-, monohydrochloride
PubChem CID 115000
IUPAC Name N-(1-azabicyclo[2.2.2]octan-3-yl)-6-chloro-4-methyl-3-oxo-1,4-benzoxazine-8-carboxamide;hydrochloride
SMILES CN1C(=O)COC2=C1C=C(C=C2C(=O)NC3CN4CCC3CC4)Cl.Cl

Specifications

Quantity 50 mg
Formula Weight 386.28
Percent Purity >99%
Chemical Name or Material Y-25130 hydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.