Learn More
Medchemexpress LLC HY-101069 2mg Medchemexpress, Y-26763 CAS:127408-31-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010692MG
Encompass_Preferred
Medchemexpress, HY-101069 2mg Y-26763 CAS:127408-31-5 Y-26763 is a K+ channel opener and active metabolite of Y-27152. Y-26763 is an ATP-sensitive K+ (KATP) channel activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7802