Learn More
Medchemexpress LLC HY-10071 10mg , Y-27632 CAS:146986-50-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007110MG
Encompass_Preferred
HY-10071 10mg Y-27632 CAS: 146986-50-7 Y-27632 is an orally active, ATP-competitive inhibitor of ROCK-I and ROCK-II, with Kis of 220 and 300 nM, respectively. Y-27632 attenuates Doxorubicin-induced apoptosis of human cardiac stem cells. Y-27632 also suppresses dissociation-induced apoptosis of murine prostate stem/progenitor cells. Y-27632 primes human induced pluripotent stem cells (hIPSCs) to selectively differentiate towards mesendodermal lineage via epithelial-mesenchymal transition-like modulation[4][5][6][7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8967