Learn More
Apexbio Technology LLC Y-27632, 146986-50-7, MFCD03093855, 10 mg
SDP
Catalog No. 501012284
Encompass_Preferred
MF: C14H21N3O, MW: 247.34, Solubility: 24.7mg/mL in DMSO,insoluble in Chloroform. Y-27632 is a specific inhibitor of Rho-associated kinases(ROCK) family with Ki values of 0.22uM and 0.30uM for ROCK1 and ROCK2, respectively.
Catalog No. 50-101-2284 Supplier Apexbio Technology LLC Supplier No. B129310
MF: C14H21N3O, MW: 247.34, Solubility: 24.7mg/mL in DMSO,insoluble in Chloroform. Y-27632 is a specific inhibitor of Rho-associated kinases(ROCK) family with Ki values of 0.22uM and 0.30uM for ROCK1 and ROCK2, respectively.