Learn More
Apexbio Technology LLC Y-27632 146986-50-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062619
Encompass_Preferred
Y-27632 (CAS 146986-50-7) is a selective inhibitor of Rho-associated protein kinases (ROCK) specifically targeting ROCK1 and ROCK2 isoforms It competitively binds to the ATP-binding site inhibiting ROCK1 and ROCK2 with Ki values of 0 22 M and 0 30 M respectively Compared to other kinases such as citron kinase PKN and PKC Y-27632 exhibits significantly higher selectivity toward ROCK isoforms In cellular assays Y-27632 at 10 M concentration effectively disrupts stress fiber formation in Swiss 3T3 fibroblast cells It is widely used in cell biology research to study ROCK signaling pathways and cytoskeletal dynamics
Catalog No. 50-000-62619 Supplier Apexbio Technology LLC Supplier No. B12935.1
Y-27632 (CAS 146986-50-7) is a selective inhibitor of Rho-associated protein kinases (ROCK) specifically targeting ROCK1 and ROCK2 isoforms It competitively binds to the ATP-binding site inhibiting ROCK1 and ROCK2 with Ki values of 0 22 M and 0 30 M respectively Compared to other kinases such as citron kinase PKN and PKC Y-27632 exhibits significantly higher selectivity toward ROCK isoforms In cellular assays Y-27632 at 10 M concentration effectively disrupts stress fiber formation in Swiss 3T3 fibroblast cells It is widely used in cell biology research to study ROCK signaling pathways and cytoskeletal dynamics