Learn More
Selleck Chemical LLC Y-27632 2HCl S1049-200mg
SDP
Catalog No. 503108351
Encompass_Preferred
Y-27632 2HCl is a selective ROCK1 and ROCK2 inhibitor with a Ki of 140 nM and 300nM in a cell-free assay exhibits 200-fold selectivity over other kinases including PKC cAMP-dependent protein kinase MLCK and PAK
Catalog No. 50-310-8351 Supplier Selleck Chemical LLC Supplier No. S1049200MG
Y-27632 2HCl is a selective ROCK1 and ROCK2 inhibitor with a Ki of 140 nM and 300nM in a cell-free assay exhibits 200-fold selectivity over other kinases including PKC cAMP-dependent protein kinase MLCK and PAK