Learn More
Apexbio Technology LLC Y-27632 dihydrochloride 129830-38-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063418
Encompass_Preferred
Y-27632 dihydrochloride (CAS 129830-38-2) is a small-molecule inhibitor targeting ROCK kinases (Rho-associated protein kinases) specifically interacting with the catalytic domains of ROCK-1 and ROCK-2 It inhibits their kinase activity with an IC50 of approximately 140 nM By targeting ROCK signaling Y-27632 disrupts Rho-mediated formation of cellular stress fibers influences cell cycle progression from G1 to S phase and interferes with cytokinesis It is commonly utilized in studies investigating cell proliferation cytoskeletal organization and stem cell viability
Catalog No. 50-000-63418 Supplier Apexbio Technology LLC Supplier No. A30085.1
Y-27632 dihydrochloride (CAS 129830-38-2) is a small-molecule inhibitor targeting ROCK kinases (Rho-associated protein kinases) specifically interacting with the catalytic domains of ROCK-1 and ROCK-2 It inhibits their kinase activity with an IC50 of approximately 140 nM By targeting ROCK signaling Y-27632 disrupts Rho-mediated formation of cellular stress fibers influences cell cycle progression from G1 to S phase and interferes with cytokinesis It is commonly utilized in studies investigating cell proliferation cytoskeletal organization and stem cell viability