Learn More
Selleck Chemical LLC Y-27632 5mg
SDP
Supplier:  Selleck Chemical LLC S63905MG
Encompass_Preferred
Y-27632 is a specific inhibitor of ROCK with an Ki of 140 nM for ROCK1 (p160ROCK). Y-27632 shows >200-fold selectivity over other kinases, including PKC, cAMP-dependent protein kinase, MLCK and PAK. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3178