Learn More
Selleck Chemical LLC Y-27632
SDP
Catalog No. 5000004653
Encompass_Preferred
Y-27632 is a selective ROCK1 and ROCK2 inhibitor with a Ki of 140 nM and 300nM in a cell-free assay exhibits 200-fold selectivity over other kinases including PKC cAMP-dependent protein kinase MLCK and PAK
Catalog No. 50-000-04653 Supplier Selleck Chemical LLC Supplier No. S639010MM/1ML
Y-27632 is a selective ROCK1 and ROCK2 inhibitor with a Ki of 140 nM and 300nM in a cell-free assay exhibits 200-fold selectivity over other kinases including PKC cAMP-dependent protein kinase MLCK and PAK