Learn More
Medchemexpress LLC Y-27632 dihydrochloride | 129830-38-2 | MFCD03490488 | 100.0% | 320.26 g·mol⁻¹ | C14H23Cl2N3O | 10 MG
SDP
Catalog No. 5000243293
Encompass_Preferred
Y-27632 dihydrochloride is an analytical standard of a Rho-associated protein kinase (ROCK) inhibitor used in biochemical and pharmacological research. Supplied as a purified solid for assay development and analytical methods, it is commonly used as a reference standard in chromatographic and mass spectrometry workflows.
  • Potent ROCK inhibitor for biochemical and pharmacological studies.
  • High purity analytical standard (≥99.99%).
  • Suitable for HPLC, GC, and MS assay development.
  • Solid form, easy to handle and store.
  • Molecular weight 320.26 g·mol⁻¹ and CAS 129830-38-2.
  • COA and SDS documentation available for quality control.
Catalog No. 50-002-43293 Supplier Medchemexpress LLC Supplier No. HY10583R10MG
Y-27632 dihydrochloride is an analytical standard of a Rho-associated protein kinase (ROCK) inhibitor used in biochemical and pharmacological research. Supplied as a purified solid for assay development and analytical methods, it is commonly used as a reference standard in chromatographic and mass spectrometry workflows.
  • Potent ROCK inhibitor for biochemical and pharmacological studies.
  • High purity analytical standard (≥99.99%).
  • Suitable for HPLC, GC, and MS assay development.
  • Solid form, easy to handle and store.
  • Molecular weight 320.26 g·mol⁻¹ and CAS 129830-38-2.
  • COA and SDS documentation available for quality control.