Learn More
Medchemexpress LLC Y-27632 dihydrochloride | 129830-38-2 | MFCD03490488 | 100.0% | 320.26 g·mol⁻¹ | C14H23Cl2N3O | 25 MG
SDP
Catalog No. 5000241921
Encompass_Preferred
Y-27632 dihydrochloride is an analytical-grade Rho-associated kinase (ROCK) inhibitor provided as the dihydrochloride salt for research use. It selectively inhibits ROCK-I and ROCK-II and is used in biochemical and cell-based assays to study signaling pathways, cytoskeletal dynamics, and pharmacological responses. High purity and small-scale packaging support assay development and reproducible results.
  • High purity (99.99%).
  • Selective ROCK-I and ROCK-II inhibition.
  • Suitable for biochemical and cell-based assays.
  • Available in small package sizes for assay development.
  • COA and SDS available for quality and safety information.
  • Molecular weight 320.26 g·mol⁻¹.
Catalog No. 50-002-41921 Supplier Medchemexpress LLC Supplier No. HY10583R25MG
Y-27632 dihydrochloride is an analytical-grade Rho-associated kinase (ROCK) inhibitor provided as the dihydrochloride salt for research use. It selectively inhibits ROCK-I and ROCK-II and is used in biochemical and cell-based assays to study signaling pathways, cytoskeletal dynamics, and pharmacological responses. High purity and small-scale packaging support assay development and reproducible results.
  • High purity (99.99%).
  • Selective ROCK-I and ROCK-II inhibition.
  • Suitable for biochemical and cell-based assays.
  • Available in small package sizes for assay development.
  • COA and SDS available for quality and safety information.
  • Molecular weight 320.26 g·mol⁻¹.