Learn More
Medchemexpress LLC Y-27632 dihydrochloride | 129830-38-2 | MFCD03490488 | 99.9% | 320.26 | C14H23Cl2N3O | 100 MG
SDP
Catalog No. 5000243729
Encompass_Preferred
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated kinases (ROCK) used as a research reagent in cell biology and pharmacology studies. It is orally active and has documented kinase selectivity and potency.
  • ATP-competitive inhibitor of ROCK-I and ROCK-II.
  • Orally active compound with reported antiepileptic effects.
  • Reported Ki values: ROCK-I 220 nM, ROCK-II 300 nM.
  • High reported purity (≈99.94%).
  • Available in multiple solid pack sizes and bulk by inquiry.
Catalog No. 50-002-43729 Supplier Medchemexpress LLC Supplier No. HY10583G100MG
Add to Cart
Add to Cart
Request a Quote
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated kinases (ROCK) used as a research reagent in cell biology and pharmacology studies. It is orally active and has documented kinase selectivity and potency.
  • ATP-competitive inhibitor of ROCK-I and ROCK-II.
  • Orally active compound with reported antiepileptic effects.
  • Reported Ki values: ROCK-I 220 nM, ROCK-II 300 nM.
  • High reported purity (≈99.94%).
  • Available in multiple solid pack sizes and bulk by inquiry.