Learn More
Medchemexpress LLC Y-27632 dihydrochloride | 129830-38-2 | MFCD03490488 | 99.9% | 320.26 g/mol | C14H23Cl2N3O | 5 MG
SDP
Catalog No. 5000243728
Encompass_Preferred
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated protein kinases (ROCK) used in research to modulate cytoskeletal dynamics, cell morphology, and motility in vitro and in vivo. It is supplied as a high-purity, white to off-white solid suitable for laboratory assays and preclinical studies.
  • ATP-competitive ROCK inhibitor (ROCK-I Ki = 220 nM; ROCK-II Ki = 300 nM).
  • Orally active; applicable to in vivo and in vitro studies.
  • High purity (≈99.9%) for reproducible experimental results.
  • Molecular weight 320.26 g/mol for stoichiometric calculations.
  • White to off-white solid; convenient handling and storage.
  • Available in small research quantities suitable for assay development.
Catalog No. 50-002-43728 Supplier Medchemexpress LLC Supplier No. HY105835MG
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated protein kinases (ROCK) used in research to modulate cytoskeletal dynamics, cell morphology, and motility in vitro and in vivo. It is supplied as a high-purity, white to off-white solid suitable for laboratory assays and preclinical studies.
  • ATP-competitive ROCK inhibitor (ROCK-I Ki = 220 nM; ROCK-II Ki = 300 nM).
  • Orally active; applicable to in vivo and in vitro studies.
  • High purity (≈99.9%) for reproducible experimental results.
  • Molecular weight 320.26 g/mol for stoichiometric calculations.
  • White to off-white solid; convenient handling and storage.
  • Available in small research quantities suitable for assay development.