Learn More
Medchemexpress LLC Y-27632 dihydrochloride | 129830-38-2 | MFCD03490488 | 99.9% | 320.26 g/mol | C14H23Cl2N3O | 50 MG
SDP
Catalog No. 5000243730
Encompass_Preferred
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated protein kinases (ROCK I and II) used in cellular and pharmacological research to modulate ROCK signaling. Supplied as a high-purity solid, it is commonly used in cell signaling studies, pharmacodynamic assays, and stem cell culture applications that require ROCK pathway inhibition.
  • ATP-competitive inhibitor of ROCK I and II with reported activity in the low hundreds of nanomolar range.
  • High purity suitable for research applications.
  • Solid form for easy handling and storage.
  • Available in research-scale package sizes for laboratory use.
  • Applicable to cell signaling, pharmacology, and stem cell culture studies.
Catalog No. 50-002-43730 Supplier Medchemexpress LLC Supplier No. HY10583G50MG
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated protein kinases (ROCK I and II) used in cellular and pharmacological research to modulate ROCK signaling. Supplied as a high-purity solid, it is commonly used in cell signaling studies, pharmacodynamic assays, and stem cell culture applications that require ROCK pathway inhibition.
  • ATP-competitive inhibitor of ROCK I and II with reported activity in the low hundreds of nanomolar range.
  • High purity suitable for research applications.
  • Solid form for easy handling and storage.
  • Available in research-scale package sizes for laboratory use.
  • Applicable to cell signaling, pharmacology, and stem cell culture studies.