Learn More
Apexbio Technology LLC Y-27632 dihydrochloride, 146986-50-7, MFCD03490488, 50 mg
SDP
Supplier:  Apexbio Technology LLC A300850
Encompass_Preferred
MF: C14H21N3O2HCl, MW: 320.26, Solubility: 52.9mg/ml in H2O; 16.013mg/mL in DMSO. Y-27632 dihydrochloride is a small-molecule inhibitor of Rho-associated protein kinase p160ROCK with the IC50 of 140 nM. Y-27632 suppresses the kinase activity of both ROCK-1 and ROCK-2 in vitro, and this compound inhibits the kinases by binding to the catalytic site of ROCK-1 and ROCK-2. Thus Y-27632 function on Rho-mediated stress fiber formation, the G1-S phase progression and cytokinesis.
Catalog No. 50-101-2282