Learn More
FUJIFILM BIOSCIENCES INC Y-27632 GMP-compliant
Catalog No. 5000450295
Encompass_Preferred
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It enhances cell viability and reduces cell death during passaging and cryopreservation It is a selective and potent inhibitor of ROCK Rho-associated coiled-coil forming kinase/Rho binding kinase Y-27632 has a variety of activities including vascular smooth muscle contraction through the ROCK signaling Ki 140 nmol/L p160ROCK This product has been reported to suppress death of human ES/iPS cells at the time of cell dispersion and improves viability of the cells after cryopreservation This product complies with ICH-Q7 GMP for APIs as a raw material for commercial production of regenerative medicine products
Catalog No. 50-004-50295 Supplier FUJIFILM BIOSCIENCES INC Supplier No. 25800703
Add to Cart
Add to Cart
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It enhances cell viability and reduces cell death during passaging and cryopreservation It is a selective and potent inhibitor of ROCK Rho-associated coiled-coil forming kinase/Rho binding kinase Y-27632 has a variety of activities including vascular smooth muscle contraction through the ROCK signaling Ki 140 nmol/L p160ROCK This product has been reported to suppress death of human ES/iPS cells at the time of cell dispersion and improves viability of the cells after cryopreservation This product complies with ICH-Q7 GMP for APIs as a raw material for commercial production of regenerative medicine products