Learn More
Cayman Chemical Y-27632 (hydrochloride), 129830-38-2, 10 mg
SDP
Supplier:  Cayman Chemical 1000558310
Encompass_Preferred
A potent, ATP-competitive inhibitor of ROCKs including p160ROCK (Ki = 140 nM) and ROCK-II (IC50 = 800 nM); also inhibits PRK2 with an IC50 value of 600 nM
Catalog No. NC0140250