Learn More
FUJIFILM Biosciences Y-27632 MF
Catalog No. 5000450296
Encompass_Preferred
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It is a selective and potent inhibitor of ROCK Rho-associated coiled-coil forming kinase/Rho binding kinase Y-27632 has a variety of activities including vascular smooth muscle contraction through the ROCK signaling Ki 140 nmol/L p160ROCK This product has been reported to suppress death of human ES cells and human iPS cells at the time of cell dispersion and also improve viability of the cells after cryopreservation
Catalog No. 50-004-50296 Supplier FUJIFILM Biosciences Supplier No. 25900613
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It is a selective and potent inhibitor of ROCK Rho-associated coiled-coil forming kinase/Rho binding kinase Y-27632 has a variety of activities including vascular smooth muscle contraction through the ROCK signaling Ki 140 nmol/L p160ROCK This product has been reported to suppress death of human ES cells and human iPS cells at the time of cell dispersion and also improve viability of the cells after cryopreservation