Learn More
Medchemexpress LLC Y-320 | 288250-47-5 | 99.0% | 505.01 | 10 MG
SDP
Catalog No. 5000365132
Encompass_Preferred
Y-320 is a potent, orally active phenylpyrazoleanilide immunomodulator. It inhibits IL-17 production by CD4 T cells stimulated with IL-15 and enhances TP53, DMD, and COL17A1 PTC readthrough by G418. This compound significantly sensitizes multidrug resistance tumors by inducing G2/M phase arrest and apoptosis when used with low-dose Paclitaxel, making it suitable for research on rheumatoid arthritis and cancer.
  • Potent, orally active immunomodulator
  • Inhibits IL-17 production by CD4 T cells
  • Enhances PTC readthrough for TP53, DMD, and COL17A1
  • Increases cellular protein levels and synthesis
  • Sensitizes multidrug resistance (MDR) tumors
  • Induces G2/M phase arrest and apoptosis
  • Ameliorates collagen-induced arthritis (CIA)
  • Used in research for rheumatoid arthritis and cancer
Catalog No. 50-003-65132 Supplier Medchemexpress LLC Supplier No. 10MGHY15898
Y-320 is a potent, orally active phenylpyrazoleanilide immunomodulator. It inhibits IL-17 production by CD4 T cells stimulated with IL-15 and enhances TP53, DMD, and COL17A1 PTC readthrough by G418. This compound significantly sensitizes multidrug resistance tumors by inducing G2/M phase arrest and apoptosis when used with low-dose Paclitaxel, making it suitable for research on rheumatoid arthritis and cancer.
  • Potent, orally active immunomodulator
  • Inhibits IL-17 production by CD4 T cells
  • Enhances PTC readthrough for TP53, DMD, and COL17A1
  • Increases cellular protein levels and synthesis
  • Sensitizes multidrug resistance (MDR) tumors
  • Induces G2/M phase arrest and apoptosis
  • Ameliorates collagen-induced arthritis (CIA)
  • Used in research for rheumatoid arthritis and cancer