Learn More
Selleck Chemical LLC Y-320
SDP
Catalog No. 5000001510
Encompass_Preferred
Y-320 is an orally active immunomodulator and inhibits IL-17 production by CD4 T cells stimulated with IL-15 with IC50 of 20 to 60 nM
Catalog No. 50-000-01510 Supplier Selleck Chemical LLC Supplier No. S751610MG
Y-320 is an orally active immunomodulator and inhibits IL-17 production by CD4 T cells stimulated with IL-15 with IC50 of 20 to 60 nM