Learn More
Medchemexpress LLC HY-10067 10mg Medchemexpress, Y-33075 CAS:199433-58-4 Purity:>98%
SDP
Catalog No. 501931912
Encompass_Preferred
Medchemexpress, HY-10067 10mg Y-33075 CAS:199433-58-4 Y-33075 is a selective ROCK inhibitor derived from Y-27632, and is more potent than Y-27632, with an IC 50 of 3.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1912 Supplier Medchemexpress LLC Supplier No. HY1006710MG
Add to Cart
Add to Cart
Medchemexpress, HY-10067 10mg Y-33075 CAS:199433-58-4 Y-33075 is a selective ROCK inhibitor derived from Y-27632, and is more potent than Y-27632, with an IC 50 of 3.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.