Learn More
Medchemexpress LLC HY-10069 5mg Medchemexpress, Y-33075 (dihydrochloride) CAS:173897-44-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100695MG
Encompass_Preferred
Medchemexpress, HY-10069 5mg Y-33075 (dihydrochloride) CAS:173897-44-4 Y-33075 dihydrochloride is a selective ROCK inhibitor with an IC 50 of 3.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1918