Learn More
TARGETMOL CHEMICALS INC Y-33075 dihydrochloride 10MG
SDP
Catalog No. 502878739
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Y-33075 dihydrochloride is a selective inhibitor of ROCK with an IC50 of 3.6 nM. Purity 99.43%
Catalog No. 50-287-8739 Supplier TARGETMOL CHEMICALS INC Supplier No. T13384L10MG
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Y-33075 dihydrochloride is a selective inhibitor of ROCK with an IC50 of 3.6 nM. Purity 99.43%