Learn More
Medchemexpress LLC Y-33075 dihydrochloride | 173897-44-4 | 98.0% | 353.25 | C16H18Cl2N4O | 100 MG
SDP
Catalog No. 5000214529
Encompass_Preferred
A potent, selective Rho-associated protein kinase (ROCK) inhibitor supplied as the dihydrochloride salt for research use, suitable for cellular and biochemical assays.
  • Selective Rho-associated protein kinase (ROCK) inhibition with an IC50 of 3.6 nM.
  • High purity (98.02%), appropriate for preclinical research applications.
  • Well-characterized chemical identity: C16H18Cl2N4O, MW 353.25, CAS 173897-44-4.
  • Available in multiple pack sizes, including 100 MG, for experimental flexibility.
  • Accompanied by datasheet and batch-specific certificate of analysis for quality assurance.
Catalog No. 50-002-14529 Supplier Medchemexpress LLC Supplier No. HY10069100MG
A potent, selective Rho-associated protein kinase (ROCK) inhibitor supplied as the dihydrochloride salt for research use, suitable for cellular and biochemical assays.
  • Selective Rho-associated protein kinase (ROCK) inhibition with an IC50 of 3.6 nM.
  • High purity (98.02%), appropriate for preclinical research applications.
  • Well-characterized chemical identity: C16H18Cl2N4O, MW 353.25, CAS 173897-44-4.
  • Available in multiple pack sizes, including 100 MG, for experimental flexibility.
  • Accompanied by datasheet and batch-specific certificate of analysis for quality assurance.