Learn More
Medchemexpress LLC Y-33075 dihydrochloride | 173897-44-4 | 98.8% | 353.25 | 50 MG
SDP
Catalog No. 5000365711
Encompass_Preferred
Y-33075 dihydrochloride is a selective ROCK inhibitor with an IC50 of 3.6 nM. It also inhibits PKC and CaMKII, and has shown effects in extending neurites in retinal ganglion cells and lowering intraocular pressure.
  • Potent and selective ROCK inhibitor
  • Inhibits PKC and CaMKII
  • Extends neurites in retinal ganglion cells
  • Lowers intraocular pressure
  • Increases regenerating axons
Catalog No. 50-003-65711 Supplier Medchemexpress LLC Supplier No. 50MGHY10069
Y-33075 dihydrochloride is a selective ROCK inhibitor with an IC50 of 3.6 nM. It also inhibits PKC and CaMKII, and has shown effects in extending neurites in retinal ganglion cells and lowering intraocular pressure.
  • Potent and selective ROCK inhibitor
  • Inhibits PKC and CaMKII
  • Extends neurites in retinal ganglion cells
  • Lowers intraocular pressure
  • Increases regenerating axons