Learn More
Medchemexpress LLC Y-33075 dihydrochloride | 173897-44-4 | 98.81% | 353.25 | 25 MG
SDP
Catalog No. 5000347724
Encompass_Preferred
Y-33075 dihydrochloride is a selective ROCK inhibitor with an IC50 of 3.6 nM.
  • Selective ROCK inhibitor with an IC50 of 3.6 nM.
  • Inhibits PKC (IC50 of 420 nM) and CaMKII (IC50 of 810 nM) more potently than Y-27632.
  • Extends neurites in retinal ganglion cells (RGCs).
  • Inhibits the contraction of rabbit ciliary artery segments evoked by histamine in Ca2+-free solutions.
  • Significantly lowers intraocular pressure (IOP) in rabbits and monkeys after topical administration.
  • Increases regenerating axons of retinal ganglion cells (RGCs) in rats.
Catalog No. 50-003-47724 Supplier Medchemexpress LLC Supplier No. 25MGHY10069
Y-33075 dihydrochloride is a selective ROCK inhibitor with an IC50 of 3.6 nM.
  • Selective ROCK inhibitor with an IC50 of 3.6 nM.
  • Inhibits PKC (IC50 of 420 nM) and CaMKII (IC50 of 810 nM) more potently than Y-27632.
  • Extends neurites in retinal ganglion cells (RGCs).
  • Inhibits the contraction of rabbit ciliary artery segments evoked by histamine in Ca2+-free solutions.
  • Significantly lowers intraocular pressure (IOP) in rabbits and monkeys after topical administration.
  • Increases regenerating axons of retinal ganglion cells (RGCs) in rats.