Learn More
Selleck Chemical LLC Y-39983 Dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S7935100MG
Encompass_Preferred
Y-39983 (Y-33075)Dihydrochloride is a selective rho-associated protein kinase(ROCK) inhibitor with an IC50 of 3 6 nM
Catalog No. 50-000-01250