Learn More
Bellco Glass, Inc. Y Barb Tubing Connector,1/4ID Pkg of 10"
SDP
Catalog No. 501946380
Encompass_Preferred
Y Barbed Tubing Connector ¼". Y connector for gas blending standsMaterial: clear polycarbonate (meets fda, food & beverage). For 1/4" id silicone tubing. Max. Pressure: 125psi @72°f. Temperature range: -211° to +275°f. Use with polyurethane & pvc with a durometer of a45-a90. Single barbed connector. Technical:Polycarbonate is clear (transparent) in color to allow you to see fluids flowing through the fitting. It has better chemical resistance than nylon but si not as strong. Sterilization: steam (autoclave) not rated for vacuum.CASE/QTY: 10. MADE IN THE USA
Catalog No. 50-194-6380 Supplier Bellco Glass, Inc. Supplier No. 780800001
Y Barbed Tubing Connector ¼". Y connector for gas blending standsMaterial: clear polycarbonate (meets fda, food & beverage). For 1/4" id silicone tubing. Max. Pressure: 125psi @72°f. Temperature range: -211° to +275°f. Use with polyurethane & pvc with a durometer of a45-a90. Single barbed connector. Technical:Polycarbonate is clear (transparent) in color to allow you to see fluids flowing through the fitting. It has better chemical resistance than nylon but si not as strong. Sterilization: steam (autoclave) not rated for vacuum.CASE/QTY: 10. MADE IN THE USA