Learn More
Cusabio Technology LLC Y box binding protein 1
Catalog No. 5000049176
Encompass_Preferred
This kit detects YBX1 (Nuclease-sensitive element-binding protein 1) in human serum plasma tissue homogenates and cell lysates using a quantitative sandwich ELISA method With a detection range of 31 25 pg/mL to 2000 pg/mL and a sensitivity of 7 8 pg/mL it enables precise measurement of YBX1 concentrations across diverse biological samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is read at 450 nm for accurate quantification making it suitable for research applications requiring reliable detection of YBX1 protein levels in both clinical and experimental settings
Catalog No. 50-000-49176 Supplier Cusabio Technology LLC Supplier No. CSBEL026247HU
This kit detects YBX1 (Nuclease-sensitive element-binding protein 1) in human serum plasma tissue homogenates and cell lysates using a quantitative sandwich ELISA method With a detection range of 31 25 pg/mL to 2000 pg/mL and a sensitivity of 7 8 pg/mL it enables precise measurement of YBX1 concentrations across diverse biological samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is read at 450 nm for accurate quantification making it suitable for research applications requiring reliable detection of YBX1 protein levels in both clinical and experimental settings