Learn More
Julabo USA Inc Each, Y-adapter cable, dual Pt100 sensor to 1 Lemosa sockets
SDP
Catalog No. 507534247
Encompass_Preferred
Bath Accessories, Julabo,Y-adapter cable for connecting 1 dual Pt100 sensor to 2 Lemosa sockets for LC Temperature Laboratory Controller
Catalog No. 50-753-4247 Supplier Julabo USA Inc Supplier No. 8980142
Bath Accessories, Julabo,Y-adapter cable for connecting 1 dual Pt100 sensor to 2 Lemosa sockets for LC Temperature Laboratory Controller