Learn More
Thermo Scientific Y connector 0.5mm PEEK [quantity]
Supplier:  Thermo Scientific 1600583
Encompass
Y shaped fluidic connector with 0.5 mm inner diameter made of polyetheretherketone plastic splits and combines fluid flow enabling flexible microfluidics routing and is commonly used in analytical instrumentation setups.
Catalog No. NC9978556