Learn More
Instech Three way Y connector, 22ga, sterile.
SDP
Catalog No. 5081210
Encompass
The Instech SCY22 is a sterile three‑way Y connector used to combine flow paths in small‑animal infusion and glucose clamp studies. Constructed with 22ga stainless steel tubes and a chemically resistant Kel‑F hub, it provides a secure, low‑dead‑volume junction, with each tube extending approximately 6mm from the hub for easy integration with standard laboratory tubing.
Features
  • Sterile 3‑way Y connector for multi‑path fluid routing
  • 22ga stainless steel tubes with rounded edges
  • Chemically resistant Kel‑F (PCTFE) hub
  • Low dead volume of less than 3µL
  • Tubes extend ~6mm for secure tubing attachment
Catalog No. 50-812-10 Supplier Instech Supplier No. SCY22
The Instech SCY22 is a sterile three‑way Y connector used to combine flow paths in small‑animal infusion and glucose clamp studies. Constructed with 22ga stainless steel tubes and a chemically resistant Kel‑F hub, it provides a secure, low‑dead‑volume junction, with each tube extending approximately 6mm from the hub for easy integration with standard laboratory tubing.
Features
  • Sterile 3‑way Y connector for multi‑path fluid routing
  • 22ga stainless steel tubes with rounded edges
  • Chemically resistant Kel‑F (PCTFE) hub
  • Low dead volume of less than 3µL
  • Tubes extend ~6mm for secure tubing attachment