Learn More
Instech Three way Y connector, 25ga, sterile.
SDP
Catalog No. 5081211
Encompass
The Instech SCY25 is a sterile three‑way Y connector designed for combining flow paths in small‑animal infusion and glucose clamp studies. Built with 25ga stainless steel tubes and a chemically resistant Kel‑F hub, it provides an ultra‑low‑dead‑volume junction with approximately 6mm tube extensions for secure attachment to fine‑diameter laboratory tubing.
Features
  • Sterile 3‑way Y connector for multi‑path fluid routing
  • 25ga stainless steel tubes for minimal dead volume
  • Kel‑F (PCTFE) hub for chemical resistance
  • Dead volume less than 3µL
  • ~6mm tube extensions for reliable tubing engagement
Catalog No. 50-812-11 Supplier Instech Supplier No. SCY25
The Instech SCY25 is a sterile three‑way Y connector designed for combining flow paths in small‑animal infusion and glucose clamp studies. Built with 25ga stainless steel tubes and a chemically resistant Kel‑F hub, it provides an ultra‑low‑dead‑volume junction with approximately 6mm tube extensions for secure attachment to fine‑diameter laboratory tubing.
Features
  • Sterile 3‑way Y connector for multi‑path fluid routing
  • 25ga stainless steel tubes for minimal dead volume
  • Kel‑F (PCTFE) hub for chemical resistance
  • Dead volume less than 3µL
  • ~6mm tube extensions for reliable tubing engagement