Learn More
Foxx Life Sciences Polypropylene Y-Type Barbed Tubing Connectors (Wye-Shaped), Three 1/4 Inch Barbs, Autoclavable, 10/PK
Catalog No. 501178743
Encompass_Preferred
255-2601-OEM: Connector, Y Connector, Polypropylene, 1/4" Hose Barb x 1/4" Hose Barb to 1/4" Hose Barb, 10/pk. Foxx Life Sciences Y connectors are available in a variety of standard sizes. They are guaranteed to provide a secure, watertight and airtight connection. Our Y connectors are non-corrosive and durable. They are made from polypropylene.
Catalog No. 50-117-8743 Supplier Foxx Life Sciences Supplier No. 2552601OEM
255-2601-OEM: Connector, Y Connector, Polypropylene, 1/4" Hose Barb x 1/4" Hose Barb to 1/4" Hose Barb, 10/pk. Foxx Life Sciences Y connectors are available in a variety of standard sizes. They are guaranteed to provide a secure, watertight and airtight connection. Our Y connectors are non-corrosive and durable. They are made from polypropylene.