Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Y-Connector, Hose Barb Union, 1/16"" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3061443/DEL
Encompass
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
Catalog No. NC0582773
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.