Learn More
Chemglass Life Sciences Y Connector with Male Luer Lock, L/S Platinum Cured Silicone Pump Tubing, 1/8" ID x 1/4" OD Size 16
SDP
Catalog No. 5000083214
Encompass
Reduces set up time and minimizes labor cost. Y connector and male luer assembly are package 10 per bag and ready to use.Manufactured with 5' of No.16 (1/8 x 1/4') platinum cured silicone tubing.For sterile applications the assembly can be safely autoclaved.
Catalog No. 50-000-83214 Supplier Chemglass Life Sciences Supplier No. CLS139022
Reduces set up time and minimizes labor cost. Y connector and male luer assembly are package 10 per bag and ready to use.Manufactured with 5' of No.16 (1/8 x 1/4') platinum cured silicone tubing.For sterile applications the assembly can be safely autoclaved.