Learn More
Research Products International Corp Y Tubing Connectors for 1/4 Inch Inner Diameter Tubing, 12 per Package
SDP
Supplier:  Research Products International Corp F19614
Encompass
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power. Chemical-resistant polypropylene plastic is for use with a wide range of liquids or gases. FDA-grade allow for direct contact with food, drugs, and cosmetics. Fittings are autoclavable.
Catalog No. 50-998-420