Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Y-Connector, Hose Barb Union, 3/16"" ID; 100/PK
Catalog No. 50630733
Encompass
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. These nylon fittings offer good chemical resistance. Temp range: -50 to 250¿F (-46 to 121¿C). Sterilize by ethylene oxide only.
Catalog No. 50-630-733 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3072724
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. These nylon fittings offer good chemical resistance. Temp range: -50 to 250¿F (-46 to 121¿C). Sterilize by ethylene oxide only.