Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Y-Connector, Hose Barb Union, 3/8"" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3072626
Encompass
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. These nylon fittings offer good chemical resistance. Temp range: -50 to 250¿F (-46 to 121¿C). Sterilize by ethylene oxide only.
Catalog No. 50-630-774