Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, PVDF, Y-Connector, Hose Barb Union, 3/16"" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3072664
Encompass
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. The PVDF fittings offer excellent chemical resistance. Temp range: -80 to 265¿F (-62 to 129¿
Catalog No. 50-630-730