Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Y-Connector, Hose Barb Union, 1/2"" ID; 10/PK
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3061421
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. These High-Density Polyethylene (HDPE) fittings offer very good chemical resistance. Temp range: -65 to 150¿F (-53 to 65¿C). Sterilize by ethylene oxide only.