Learn More
Stoelting™ Y Maze
SDP
Catalog No. 10000329 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-329 Rat
10-000-306 Mouse
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-329 Supplier Stoelting™ Supplier No. 60280
Add to Cart
Add to Cart
Stoelting™ Y-maze is similar to the T-maze, but has three identical arms. Gradual turns of the Y-maze decrease learning time as compared to the sharp turns of the T-maze.
Specifications
For Use With (Application) Neuroscience
Animal Rat