Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Y-PER™ Plus Dialyzable Yeast Protein Extraction Reagent

Catalog No. PI78999 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78999 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI78999 Supplier Thermo Scientific™ Supplier No. 78999
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Extract and solubilize proteins for analysis from yeast and bacterial cells with this effective, fully dialyzable lysis reagent.

Effectively lyse yeast and bacterial cells to extract and solubilize proteins for analysis with Thermo Scientific™ Y-PER-Plus Dialyzable Yeast Protein Extraction Reagent.

The proprietary formulation of cell lysis buffer facilitates protein extraction from S. cerevisiae and S. pastoris yeast cell cultures. Glass bed disruption is not necessary; protein extraction is achieved by a 20-minute incubation in the reagent at room temperature. Unlike other cell lysis buffers, the Y-PER-Plus Reagent uses a detergent component that is fully dialyzable, allowing complete exchange of the extracted protein into any buffer required by downstream techniques.

Highlights:

  • Stable formulation designed for room temperature storage
  • Yields protein extract with simple 20-minute hands-off incubation
  • Requires no rigorous mechanical disruption or lengthy enzymatic treatments
  • Tris-based buffer has a very low ionic strength and does not include sodium chloride
  • Simple formulation contains no reducing agents or chelating agents
  • Fully dialyzable formulation provides complete control over downstream applications
  • Compatible with both the Thermo Scientific Pierce™ BCA and Coomassie Plus™ Protein Assays
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Format Liquid
Product Type Yeast Protein Extraction Reagent
Volume (Metric) 500 mL
Content And Storage Upon receipt store at room temperature. Product shipped at ambient temperature.
Product Line Y-PER

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.