Learn More
Bellco Glass, Inc. Y Shape CPC Connector for Inner Dia #3/8 Hose, Individually Wrapped, Sterile, 1/pk, 5/cs
SDP
Catalog No. 502330827
Encompass_Preferred
Y Shape CPC Connector for Inner Dia #3/8 Hose, Individually Wrapped, Sterile, 1/pk, 5/cs. Using with BioFactory accessories during cell culture process could simplify the liquid transfer process,meanwhile effectively reduce the risk of cell contamination. • Sterilized• Individually Wrapped in Plastic Sterile Bag
Catalog No. 50-233-0827 Supplier Bellco Glass, Inc. Supplier No. 451400113
Y Shape CPC Connector for Inner Dia #3/8 Hose, Individually Wrapped, Sterile, 1/pk, 5/cs. Using with BioFactory accessories during cell culture process could simplify the liquid transfer process,meanwhile effectively reduce the risk of cell contamination. • Sterilized• Individually Wrapped in Plastic Sterile Bag