Learn More
Hach Company Y Strainer Assembly for Hach Process Analyzers
Supplier:  Hach Company 6784800
Encompass
The Y strainer assembly is designed for process analyzers to protect internal components and sample lines by removing suspended particulates and reducing fouling.
  • Protects analyzer components by removing particulates from the sample stream.
  • 316 stainless steel construction for corrosion resistance.
  • 80 mesh screen for effective fine-particle filtration.
  • Inline Y form factor for compact installation in sample plumbing.
Catalog No. NC3361135