Learn More
Medchemexpress LLC HY-111502 10mg Medchemexpress, Y06036 CAS:1832671-96-1 Purity:>98%
SDP
Catalog No. 501968892
Encompass_Preferred
Medchemexpress, HY-111502 10mg Y06036 CAS:1832671-96-1 Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8892 Supplier Medchemexpress LLC Supplier No. HY11150210MG
Medchemexpress, HY-111502 10mg Y06036 CAS:1832671-96-1 Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.