Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Y13g dihydrochloride | 99.9% | 381.29 g/mol | C16H26Cl2N2O4 | 5 MG
SDP

Catalog No. 5000264836
Encompass_Preferred

Y13g dihydrochloride is a research-grade small molecule inhibitor of acetylcholinesterase (AChE) and interleukin-6 (IL-6), used in in vitro and in vivo pharmacology studies. It has been shown to reverse STZ-induced memory deficits in animal models. For research use only; not for human or diagnostic use.

  • High purity (99.9%)
  • Molecular formula C16H26Cl2N2O4; molecular weight 381.29 g/mol
  • Solid, off-white to light yellow
  • Soluble in DMSO (≈125 mg/mL)
  • Storage: sealed at -20°C; in solution: -80°C (6 months), -20°C (1 month)
  • Targets acetylcholinesterase and interleukin-6
  • Suitable for in vitro and in vivo studies
  • Not for human or diagnostic use

Catalog No. 50-002-64836 Supplier Medchemexpress LLC Supplier No. HY115910A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Y13g dihydrochloride is a research-grade small molecule inhibitor of acetylcholinesterase (AChE) and interleukin-6 (IL-6), used in in vitro and in vivo pharmacology studies. It has been shown to reverse STZ-induced memory deficits in animal models. For research use only; not for human or diagnostic use.

  • High purity (99.9%)
  • Molecular formula C16H26Cl2N2O4; molecular weight 381.29 g/mol
  • Solid, off-white to light yellow
  • Soluble in DMSO (≈125 mg/mL)
  • Storage: sealed at -20°C; in solution: -80°C (6 months), -20°C (1 month)
  • Targets acetylcholinesterase and interleukin-6
  • Suitable for in vitro and in vivo studies
  • Not for human or diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.