Learn More
TARGETMOL CHEMICALS INC Y15 100MG
SDP
Catalog No. 502576353
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Y15 (1245-Benzenetetramine tetrahydrochlor) is a potent and specificsmall-molecule FAK scaffolding inhibitor that inhibits its autophosphorylation activity blocks tumor growth. purity: 98%
Catalog No. 50-257-6353 Supplier TARGETMOL CHEMICALS INC Supplier No. T7119100MG
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Y15 (1245-Benzenetetramine tetrahydrochlor) is a potent and specificsmall-molecule FAK scaffolding inhibitor that inhibits its autophosphorylation activity blocks tumor growth. purity: 98%