Learn More
Medchemexpress LLC HY-12444 10mg Medchemexpress, Y15 CAS:4506-66-5 Purity:>98%
SDP
Catalog No. 502081045
Encompass_Preferred
Medchemexpress, HY-12444 10mg Y15 CAS:4506-66-5 Y15 is a potent and specific inhibitor of focal adhesion kinase (FAK) that inhibits its autophosphorylation activity, decreases the viability of cancer cells, and blocks tumor growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1045 Supplier Medchemexpress LLC Supplier No. HY1244410MG
Medchemexpress, HY-12444 10mg Y15 CAS:4506-66-5 Y15 is a potent and specific inhibitor of focal adhesion kinase (FAK) that inhibits its autophosphorylation activity, decreases the viability of cancer cells, and blocks tumor growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.